Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145838.1 | NP_665837.1 | 4 - 5 | 456 | 64 | |
GenBank mRNA | AB059554.1 | - | 4 - 5 | 456 | 64 | |
AK085105.1 | - | 5 - 6 | 593 | 64 | ||
AK156557.1 | - | 4 - 5 | 993 | 64 | ||
AK172400.1 | - | 4 - 5 | 521 | 64 | ||
BC115912.1 | - | 4 - 5 | 411 | 40104779 | 64 | |
BC118063.1 | - | 4 - 5 | 411 | 40104780 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |