Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145940.2 | NP_666052.1 | 9 - 10 | 1043 | 55 | |
GenBank mRNA | AK031672.1 | - | 9 - 10 | 1052 | 55 | |
AK034907.1 | - | 9 - 10 | 1087 | 55 | ||
AK050495.1 | - | 9 - 10 | 1051 | 55 | ||
AK159608.1 | - | 9 - 10 | 1072 | 55 | ||
AK170563.1 | - | 9 - 10 | 1087 | 55 | ||
BC024811.1 | - | 9 - 10 | 932 | 55 | ||
BC024883.1 | - | 9 - 10 | 1007 | 55 | ||
BC025560.1 | - | 9 - 10 | 1043 | 5322999 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |