Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcmbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145955.3 | NP_666067.1 | 6 - 7 | 812 | 72 | |
GenBank mRNA | AK083385.1 | - | 6 - 7 | 873 | 72 | |
BC025641.1 | - | 6 - 7 | 842 | 5322565 | 72 | |
BC038342.1 | - | 6 - 7 | 812 | 4191750 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |