Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fev | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153111.2 | NP_694751.1 | 1 - 2 | 253 | 70 | |
GenBank mRNA | AY049085.1 | - | 1 - 2 | 263 | 70 | |
BC138435.1 | - | 1 - 2 | 168 | 8861455 | 70 | |
BC138436.1 | - | 1 - 2 | 168 | 8861456 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |