Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153122.2 | NP_694762.1 | 6 - 7 | 860 | 61 | |
GenBank mRNA | AK158391.1 | - | 6 - 7 | 902 | 61 | |
BC034522.1 | - | 6 - 7 | 860 | 4236444 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |