Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oplah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153122.2 | NP_694762.1 | 18 - 19 | 2579 | 66 | |
GenBank mRNA | AK158391.1 | - | 18 - 19 | 2621 | 66 | |
AK164524.1 | - | 2 - 3 | 203 | 66 | ||
BC025120.1 | - | 5 - 6 | 677 | 4020691 | 66 | |
BC034522.1 | - | 18 - 19 | 2579 | 4236444 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |