Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153145.4 | NP_694785.3 | 10 - 11 | 1514 | 134 | |
GenBank mRNA | AB255881.1 | - | 9 - 10 | 1433 | 134 | |
AB255882.1 | - | 10 - 11 | 1521 | 134 | ||
AF498360.1 | - | 9 - 10 | 1442 | 134 | ||
AY732492.1 | - | 9 - 10 | 1442 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |