Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178058.1 | NP_001171529.1 | 5 - 6 | 762 | 88 | |
NM_001178059.1 | NP_001171530.1 | 5 - 6 | 762 | 88 | ||
NM_001178060.1 | NP_001171531.1 | 5 - 6 | 762 | 88 | ||
NM_153162.3 | NP_694802.2 | 5 - 6 | 762 | 88 | ||
GenBank mRNA | AF349659.1 | - | 5 - 6 | 671 | 88 | |
AK012699.1 | - | 5 - 6 | 755 | 88 | ||
AK041046.1 | - | 5 - 6 | 764 | 89 | ||
AK134987.1 | - | 5 - 6 | 753 | 88 | ||
BC076605.1 | - | 5 - 6 | 720 | 30628949 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |