Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204241.1 | NP_001191170.1 | 4 - 5 | 391 | 77 | |
NM_153197.4 | NP_694877.2 | 3 - 4 | 527 | 77 | ||
GenBank mRNA | AK014135.1 | - | 3 - 4 | 427 | 77 | |
AK156040.1 | - | 3 - 4 | 527 | 77 | ||
AY397674.1 | - | 3 - 4 | 338 | 77 | ||
BC034893.1 | - | 4 - 5 | 391 | 1496162 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |