Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153384.3 | NP_700433.1 | 2 - 3 | 485 | 124 | |
GenBank mRNA | AF495718.1 | - | 2 - 3 | 485 | 124 | |
AK037267.1 | - | 2 - 3 | 149 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |