Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163024.1 | NP_001156496.1 | 34 - 35 | 5444 | 79 | |
NM_001163025.1 | NP_001156497.1 | 32 - 33 | 5426 | 79 | ||
NM_153395.2 | NP_700444.2 | 32 - 33 | 5423 | 79 | ||
GenBank mRNA | AB014059.1 | - | 32 - 33 | 4945 | 79 | |
AK011826.1 | - | 11 - 12 | 2213 | 79 | ||
AK012182.2 | - | 12 - 13 | 2362 | 79 | ||
AK122425.1 | - | 34 - 35 | 5444 | 79 | ||
AK162859.1 | - | 9 - 10 | 1917 | 79 | ||
BC048406.1 | - | 21 - 22 | 3220 | 79 | ||
BC150943.1 | - | 32 - 33 | 4947 | 9087854 | 79 | |
CN702930.1 | - | 2 - 3 | 279 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |