Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Liph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083894.1 | NP_001077363.1 | 2 - 3 | 592 | 107 | |
NM_001289582.1 | NP_001276511.1 | 2 - 3 | 592 | 107 | ||
NM_153404.3 | NP_700453.1 | 2 - 3 | 592 | 107 | ||
GenBank mRNA | AK035498.1 | - | 2 - 3 | 545 | 107 | |
AK048150.1 | - | 2 - 3 | 491 | 107 | ||
AK134352.1 | - | 2 - 3 | 551 | 107 | ||
AK136503.1 | - | 2 - 3 | 555 | 107 | ||
AK161067.1 | - | 2 - 3 | 536 | 107 | ||
AK162492.1 | - | 2 - 3 | 558 | 107 | ||
AY093499.1 | - | 2 - 3 | 419 | 107 | ||
BC037489.1 | - | 2 - 3 | 510 | 4487939 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |