Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153585.5 | NP_705813.2 | 6 - 7 | 918 | 99 | |
GenBank mRNA | AK029103.1 | - | 6 - 7 | 793 | 99 | |
AK080503.1 | - | 6 - 7 | 806 | 99 | ||
AK149944.1 | - | 6 - 7 | 751 | 99 | ||
AK157824.1 | - | 6 - 7 | 817 | 99 | ||
AK161842.1 | - | 6 - 7 | 902 | 99 | ||
AK171701.1 | - | 6 - 7 | 898 | 99 | ||
BC025554.1 | - | 6 - 7 | 779 | 5321869 | 99 | |
BC036178.1 | - | 3 - 4 | 374 | 99 | ||
BC036179.1 | - | 6 - 7 | 890 | 5059723 | 99 | |
BC039183.1 | - | 6 - 7 | 892 | 2631793 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |