Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153744.3 | NP_714966.1 | 4 - 5 | 740 | 82 | |
GenBank mRNA | AF525500.1 | - | 4 - 5 | 647 | 82 | |
AF525501.1 | - | 3 - 4 | 668 | 82 | ||
AK036585.1 | - | 4 - 5 | 741 | 82 | ||
BC116749.1 | - | 3 - 4 | 604 | 40126068 | 82 | |
BC116777.1 | - | 3 - 4 | 604 | 40126096 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |