Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153744.3 | NP_714966.1 | 11 - 12 | 1306 | 71 | |
GenBank mRNA | AF525500.1 | - | 11 - 12 | 1213 | 71 | |
AF525501.1 | - | 10 - 11 | 1234 | 71 | ||
AK036585.1 | - | 11 - 12 | 1307 | 71 | ||
BC116749.1 | - | 10 - 11 | 1170 | 40126068 | 71 | |
BC116777.1 | - | 10 - 11 | 1170 | 40126096 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |