Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153775.2 | NP_722470.2 | 11 - 12 | 1305 | 56 | |
GenBank mRNA | AK012440.1 | - | 11 - 12 | 1270 | 56 | |
AK090349.1 | - | 7 - 8 | 901 | 56 | ||
AK154995.1 | - | 11 - 12 | 1240 | 56 | ||
BC036332.1 | - | 11 - 12 | 1258 | 3156706 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |