Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153781.1 | NP_722476.1 | 13 - 14 | 1690 | 68 | |
GenBank mRNA | AK032769.1 | - | 13 - 14 | 1703 | 68 | |
AK035673.1 | - | 13 - 14 | 1420 | 68 | ||
AK133794.1 | - | 4 - 5 | 420 | 68 | ||
AK134667.1 | - | 13 - 14 | 1700 | 68 | ||
AK142716.1 | - | 13 - 14 | 1700 | 68 | ||
AK147763.1 | - | 13 - 14 | 1722 | 68 | ||
AK168381.1 | - | 13 - 14 | 1702 | 68 | ||
AK169052.1 | - | 13 - 14 | 1725 | 68 | ||
AK169631.1 | - | 13 - 14 | 1704 | 68 | ||
BC032209.1 | - | 7 - 8 | 845 | 68 | ||
BC035283.1 | - | 13 - 14 | 1690 | 4947455 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |