Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc64b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153784.2 | NP_722479.1 | 3 - 4 | 121 | ||
GenBank mRNA | AK036793.1 | - | 3 - 4 | 121 | ||
AK133729.1 | - | 3 - 4 | 121 | |||
BC038613.1 | - | 3 - 4 | 4458084 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |