Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153789.3 | NP_722484.2 | 1 - 2 | 304 | 70 | |
GenBank mRNA | AK034887.1 | - | 1 - 2 | 304 | 70 | |
AK159478.1 | - | 1 - 2 | 305 | 70 | ||
AK171888.1 | - | 1 - 2 | 305 | 70 | ||
AY434449.1 | - | 1 - 2 | 89 | 70 | ||
BC010206.1 | - | 1 - 2 | 160 | 3964381 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |