Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153795.2 | NP_722490.1 | 12 - 13 | 1755 | 59 | |
GenBank mRNA | AK090332.1 | - | 12 - 13 | 1652 | 59 | |
AK142912.1 | - | 12 - 13 | 1756 | 59 | ||
AK158357.1 | - | 12 - 13 | 1652 | 59 | ||
AK169682.1 | - | 12 - 13 | 1666 | 59 | ||
BC025119.1 | - | 12 - 13 | 1687 | 4190760 | 59 | |
BC032204.1 | - | 12 - 13 | 1731 | 5342463 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |