Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC030417.1 | - | 9 - 10 | 850 | 101 | |
BC049264.1 | - | 9 - 10 | 839 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |