Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nupl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170591.1 | NP_733479.1 | 9 - 10 | 1102 | 72 | |
GenBank mRNA | AK006062.1 | - | 9 - 10 | 1205 | 72 | |
AK029688.1 | - | 1156 | 72 | |||
AK078279.1 | - | 9 - 10 | 1150 | 72 | ||
AK086412.1 | - | 9 - 10 | 1170 | 72 | ||
AK129134.1 | - | 9 - 10 | 1100 | 72 | ||
AK133721.1 | - | 9 - 10 | 1113 | 72 | ||
AK139895.1 | - | 9 - 10 | 1233 | 72 | ||
AK146477.1 | - | 9 - 10 | 1102 | 72 | ||
BC026743.1 | - | 9 - 10 | 1133 | 3964978 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |