Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163307.1 | NP_001156779.1 | 2 - 3 | 246 | 61 | |
NM_009545.2 | NP_033571.1 | 4 - 5 | 513 | 61 | ||
GenBank mRNA | BC016419.1 | - | 2 - 3 | 200 | 4458285 | 61 |
D90085.1 | - | 4 - 5 | 361 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |