Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163307.1 | NP_001156779.1 | 3 - 4 | 410 | 77 | |
NM_001163308.1 | NP_001156780.1 | 2 - 3 | 272 | 77 | ||
NM_009545.2 | NP_033571.1 | 5 - 6 | 677 | 77 | ||
GenBank mRNA | AF483502.1 | - | 1 - 2 | 118 | 77 | |
AF483503.1 | - | 1 - 2 | 118 | 77 | ||
AK142837.1 | - | 2 - 3 | 185 | 77 | ||
BC016419.1 | - | 3 - 4 | 364 | 4458285 | 77 | |
BI991588.1 | - | 2 - 3 | 338 | 77 | ||
D90085.1 | - | 5 - 6 | 525 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |