Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 2 - 3 | 574 | 106 | |
NM_009765.3 | NP_033895.2 | 2 - 3 | 380 | 106 | ||
GenBank mRNA | AK087379.1 | - | 2 - 3 | 566 | 106 | |
AK145017.1 | - | 2 - 3 | 541 | 106 | ||
AK148272.1 | - | 2 - 3 | 378 | 106 | ||
AK164598.1 | - | 2 - 3 | 370 | 106 | ||
U65594.1 | - | 2 - 3 | 357 | 106 | ||
U82270.1 | - | 1 - 2 | 66 | 106 | ||
U89652.1 | - | 1 - 2 | 104 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Mouse Androgen Panel 1700 Inventoried Probe spans exons |