Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136057.2 | NP_001129529.1 | 8 - 9 | 1066 | 118 | |
NM_001146183.1 | NP_001139655.1 | 8 - 9 | 1043 | 118 | ||
NM_009947.3 | NP_034077.1 | 8 - 9 | 869 | 118 | ||
GenBank mRNA | AB008893.1 | - | 8 - 9 | 869 | 118 | |
AK134548.1 | - | 8 - 9 | 888 | 118 | ||
AK144410.1 | - | 8 - 9 | 895 | 118 | ||
AK158638.1 | - | 6 - 7 | 2459 | 118 | ||
BC050766.1 | - | 8 - 9 | 891 | 6591063 | 118 | |
BC051994.1 | - | 7 - 8 | 1451 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |