Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028979.2 | NP_083255.1 | 1 - 2 | 347 | 63 | |
GenBank mRNA | AF336850.1 | - | 1 - 2 | 347 | 63 | |
AK018422.1 | - | 1 - 2 | 290 | 63 | ||
AK033961.1 | - | 1 - 2 | 317 | 63 | ||
AK037031.1 | - | 1 - 2 | 443 | 63 | ||
BC119556.2 | - | 1 - 2 | 271 | 40057469 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |