Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028979.2 | NP_083255.1 | 4 - 5 | 824 | 82 | |
GenBank mRNA | AF336850.1 | - | 4 - 5 | 824 | 82 | |
AK018422.1 | - | 4 - 5 | 767 | 82 | ||
AK033961.1 | - | 4 - 5 | 794 | 82 | ||
AK037031.1 | - | 4 - 5 | 920 | 82 | ||
BC119556.2 | - | 4 - 5 | 748 | 40057469 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection P450 Mus musculus Inventoried Probe spans exons |