Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080849.3 | NP_543125.1 | 7 - 8 | 1151 | 113 | |
GenBank mRNA | AF407579.1 | - | 7 - 8 | 1101 | 113 | |
AK014546.1 | - | 5 - 6 | 1327 | 113 | ||
AK154358.1 | - | 7 - 8 | 1114 | 113 | ||
BC057995.1 | - | 6 - 7 | 1233 | 113 | ||
BC070457.1 | - | 7 - 8 | 1100 | 30603080 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |