Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145537.2 | NP_663512.2 | 2 - 3 | 348 | 86 | |
GenBank mRNA | AK029164.1 | - | 2 - 3 | 299 | 86 | |
AK075650.1 | - | 2 - 3 | 262 | 86 | ||
AK079296.1 | - | 2 - 3 | 284 | 86 | ||
AK080636.1 | - | 2 - 3 | 293 | 86 | ||
AK149892.1 | - | 2 - 3 | 284 | 86 | ||
AK155024.1 | - | 2 - 3 | 311 | 86 | ||
AK166998.1 | - | 2 - 3 | 284 | 86 | ||
BC008268.1 | - | 2 - 3 | 259 | 2811074 | 86 | |
BY289191.1 | - | 2 - 3 | 349 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |