Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153166.2 | NP_694806.1 | 4 - 5 | 366 | 82 | |
GenBank mRNA | AK044042.1 | - | 4 - 5 | 292 | 82 | |
BC036971.1 | - | 4 - 5 | 377 | 4527212 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |