Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153168.2 | NP_694808.1 | 12 - 13 | 1703 | 63 | |
GenBank mRNA | AJ428066.1 | - | 12 - 13 | 1703 | 63 | |
AK032695.1 | - | 12 - 13 | 1708 | 63 | ||
AK037012.1 | - | 12 - 13 | 1711 | 63 | ||
AK045640.1 | - | 12 - 13 | 1662 | 63 | ||
AK053667.1 | - | 12 - 13 | 1698 | 63 | ||
BC053916.1 | - | 13 - 14 | 1579 | 63 | ||
BC080303.1 | - | 12 - 13 | 1590 | 5695617 | 63 | |
BC111892.1 | - | 12 - 13 | 1674 | 40060998 | 63 | |
BC112412.1 | - | 12 - 13 | 1672 | 40060997 | 63 | |
BC138206.1 | - | 12 - 13 | 1677 | 8861225 | 63 | |
BC145982.1 | - | 12 - 13 | 1677 | 40131285 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |