Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170673.3 | NP_733773.2 | 18 - 19 | 1514 | 75 | |
GenBank mRNA | AK042692.1 | - | 18 - 19 | 1515 | 75 | |
AK044080.1 | - | 18 - 19 | 1480 | 75 | ||
AK047571.1 | - | 16 - 17 | 2529 | 75 | ||
AK137422.1 | - | 18 - 19 | 1515 | 75 | ||
AK220550.1 | - | 10 - 11 | 2878 | 75 | ||
BQ550677.1 | - | 3 - 4 | 75 | |||
BQ550678.1 | - | 2 - 3 | 149 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |