Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700002285.mm.1 |
TC0700000298.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002285.mm.1 |
NM_001127324.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002285.mm.1 |
NM_001127324.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002285.mm.1 |
NM_007948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002285.mm.1 |
NM_007948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002290.mm.1 |
TC0700000298.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002290.mm.1 |
NM_001127324.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002290.mm.1 |
NM_001127324.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002290.mm.1 |
NM_007948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002290.mm.1 |
NM_007948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002287.mm.1 |
TC0700000298.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002287.mm.1 |
NM_001127324.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002287.mm.1 |
NM_001127324.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002287.mm.1 |
NM_007948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002287.mm.1 |
NM_007948.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002289.mm.1 |
TC0700000298.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002289.mm.1 |
NM_001127324.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002289.mm.1 |
NM_001127324.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002289.mm.1 |
NM_007948.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002289.mm.1 |
NM_007948.2 |
Refseq |