Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163394.1 | NP_001156866.1 | 9 - 10 | 1438 | 78 | |
NM_001163395.1 | NP_001156867.1 | 9 - 10 | 1190 | 78 | ||
NM_001163396.2 | NP_001156868.1 | 10 - 11 | 1212 | 78 | ||
NM_007965.3 | NP_031991.3 | 9 - 10 | 1438 | 78 | ||
GenBank mRNA | AF279662.1 | - | 8 - 9 | 949 | 78 | |
AK134563.1 | - | 9 - 10 | 1191 | 78 | ||
AK137823.1 | - | 9 - 10 | 1028 | 78 | ||
AK204923.1 | - | 2 - 3 | 92 | 78 | ||
BC059810.1 | - | 9 - 10 | 1438 | 6828523 | 78 | |
BC096017.1 | - | 9 - 10 | 1331 | 78 | ||
U72519.1 | - | 9 - 10 | 1201 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |