Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB004817.1 | - | 17 - 18 | 2048 | 78 | |
AF483490.1 | - | 15 - 16 | 1937 | 78 | ||
AF483491.1 | - | 15 - 16 | 1937 | 78 | ||
AK129004.1 | - | 16 - 17 | 2008 | 78 | ||
AK138942.1 | - | 17 - 18 | 2031 | 78 | ||
AK140694.1 | - | 17 - 18 | 2055 | 78 | ||
AK154565.1 | - | 17 - 18 | 2028 | 78 | ||
AK185194.1 | - | 1 - 2 | 60 | 78 | ||
BC007135.1 | - | 16 - 17 | 2008 | 3600884 | 78 | |
U60453.1 | - | 17 - 18 | 2036 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |