Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008225.2 | NP_032251.2 | 7 - 8 | 661 | 82 | |
GenBank mRNA | AK088723.1 | - | 7 - 8 | 643 | 82 | |
BC007469.1 | - | 7 - 8 | 631 | 3593456 | 82 | |
D42120.1 | - | 7 - 8 | 592 | 82 | ||
X84797.1 | - | 7 - 8 | 629 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |