Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK032572.1 | - | 9 - 10 | 2747 | 68 | |
AK143616.1 | - | 13 - 14 | 1678 | 68 | ||
AK144669.1 | - | 13 - 14 | 1685 | 68 | ||
AK146813.1 | - | 13 - 14 | 1647 | 68 | ||
AK146918.1 | - | 13 - 14 | 1655 | 68 | ||
BC003741.1 | - | 13 - 14 | 1644 | 3488803 | 68 | |
D63850.1 | - | 13 - 14 | 1661 | 68 | ||
FN687734.2 | - | 13 - 14 | 1404 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |