Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28184739 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28184741 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28184740 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28184742 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103856 |
BC003239.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103856 |
NM_008244.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103856 |
NM_001159328.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103857 |
BC003239.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103857 |
AK046299.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103857 |
NM_008244.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2103857 |
NM_001159328.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
395424 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
811229 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
31084 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
800707 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
928161 |
AB006329.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
928161 |
AJ000740.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
928161 |
BC007130.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
928161 |
NM_011439.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
604786 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453790 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858869 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
201008 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
874003 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
27513 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135142 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168831 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103856 |
BC003239.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103856 |
NM_008244.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103856 |
NM_001159328.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103857 |
BC003239.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103857 |
AK046299.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103857 |
NM_008244.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2103857 |
NM_001159328.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P460633 |
BC003239.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P460633 |
D50050.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P460633 |
AK145031.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P460633 |
AK166441.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P460633 |
AK046299.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184768 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184781 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184834 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184751 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184829 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184844 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184758 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184765 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184836 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184767 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184766 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184843 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184744 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184821 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184746 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184755 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184763 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184743 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184823 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184848 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184838 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184754 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184778 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184775 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184826 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184830 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184839 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184841 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184760 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184842 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184759 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184827 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184742 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184783 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184832 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184772 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184828 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184740 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184774 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184833 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184822 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184847 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184761 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184849 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184749 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184846 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184770 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28184752 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07516420 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135142 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168831 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02446504 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P460633 |
BC003239.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P460633 |
D50050.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P460633 |
AK145031.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P460633 |
AK166441.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P460633 |
AK046299.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133197 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168831 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135142 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09302374 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09186868 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09302372 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09302371 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28184739 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28184740 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28184742 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28184741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184918 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184896 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184901 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184803 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184882 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184877 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184912 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184892 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184898 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184864 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184894 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28184807 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184755 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184823 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184763 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184783 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184844 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184787 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09186874 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184803 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184838 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184760 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09700581 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184761 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09186879 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09302379 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184742 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184812 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184790 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09397163 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184772 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09186873 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184843 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09160059 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09186868 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184744 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184770 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184830 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184826 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09457562 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08976692 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184849 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184839 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184766 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09338746 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184836 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184817 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184765 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09063337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184833 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184778 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184752 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09160058 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09700588 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09160072 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184828 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09283910 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184832 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184807 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09338752 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184822 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09186875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184754 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09397156 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184774 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184767 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184813 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28184758 |
- |
- |