Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC005438.1 | - | 5 - 6 | 1655 | 3487600 | 124 |
BC031543.1 | - | 5 - 6 | 1381 | 3669513 | 124 | |
U28728.1 | - | 5 - 6 | 1610 | 124 | ||
U57686.1 | - | 5 - 6 | 1310 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |