Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010135.3 | NP_034265.2 | 4 - 5 | 855 | 71 | |
GenBank mRNA | U72523.1 | - | 4 - 5 | 548 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |