Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083120.2 | NP_001076589.1 | 12 - 13 | 2707 | 66 | |
NM_001083121.2 | NP_001076590.1 | 12 - 13 | 1975 | 66 | ||
NM_008680.3 | NP_032706.2 | 13 - 14 | 2719 | 66 | ||
NM_010135.3 | NP_034265.2 | 13 - 14 | 2764 | 66 | ||
GenBank mRNA | BC062927.1 | - | 12 - 13 | 1594 | 6851217 | 66 |
U72520.1 | - | 12 - 13 | 1668 | 66 | ||
U72521.1 | - | 12 - 13 | 2394 | 66 | ||
U72522.1 | - | 13 - 14 | 2406 | 66 | ||
U72523.1 | - | 13 - 14 | 2451 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |