Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500011471.mm.1 |
NM_010162.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011471.mm.1 |
NM_010162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011471.mm.1 |
TC1500001511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011477.mm.1 |
NM_010162.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011477.mm.1 |
NM_010162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011477.mm.1 |
TC1500001511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011474.mm.1 |
NM_010162.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011474.mm.1 |
NM_010162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011474.mm.1 |
TC1500001511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011475.mm.1 |
NM_010162.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011475.mm.1 |
NM_010162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011475.mm.1 |
TC1500001511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500011476.mm.1 |
NM_010162.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011476.mm.1 |
NM_010162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500011476.mm.1 |
TC1500001511.mm.1 |
MTA TC |