Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010404.3 | NP_034534.1 | 4 - 5 | 828 | 87 | |
NM_177981.2 | NP_817090.1 | 4 - 5 | 828 | 87 | ||
GenBank mRNA | AJ000262.1 | - | 4 - 5 | 774 | 87 | |
AJ002271.1 | - | 4 - 5 | 774 | 87 | ||
AK138436.1 | - | 775 | 88 | |||
BC034089.1 | - | 4 - 5 | 766 | 4481735 | 87 | |
BC053043.1 | - | 4 - 5 | 815 | 5694628 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |