Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113331.2 | NP_001106802.1 | 1 - 2 | 726 | 82 | |
NM_011368.5 | NP_035498.2 | 2 - 3 | 328 | 82 | ||
GenBank mRNA | AK049357.1 | - | 2 - 3 | 301 | 82 | |
AK155158.1 | - | 1 - 2 | 721 | 82 | ||
BC036172.1 | - | 2 - 3 | 279 | 4988907 | 82 | |
U15784.1 | - | 2 - 3 | 218 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |