Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ern2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012016.2 | NP_036146.2 | 7 - 8 | 651 | 87 | |
GenBank mRNA | AF071777.1 | - | 7 - 8 | 610 | 87 | |
AK139291.1 | - | 7 - 8 | 643 | 87 | ||
AK153659.1 | - | 7 - 8 | 641 | 87 | ||
BC108999.2 | - | 7 - 8 | 610 | 40048029 | 87 | |
BC109000.1 | - | 7 - 8 | 610 | 40048030 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |