Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013547.3 | NP_038575.2 | 5 - 6 | 510 | 65 | |
NR_027778.1 | 4 - 5 | 421 | 65 | |||
GenBank mRNA | BC024369.1 | - | 4 - 5 | 324 | 4974534 | 65 |
BC037628.1 | - | 5 - 6 | 424 | 4973560 | 65 | |
BC044758.2 | - | 4 - 5 | 310 | 5099956 | 65 | |
BC055029.1 | - | 5 - 6 | 391 | 5054479 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |