Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013552.2 | NP_038580.2 | 10 - 11 | 1123 | 133 | |
GenBank mRNA | AF031932.1 | - | 10 - 11 | 1108 | 133 | |
AF079222.1 | - | 11 - 12 | 1072 | 133 | ||
AK143123.1 | - | 10 - 11 | 1123 | 133 | ||
AK163410.1 | - | 10 - 11 | 1066 | 133 | ||
BC021427.1 | - | 10 - 11 | 1121 | 5035341 | 133 | |
X64550.1 | - | 6 - 7 | 736 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |