Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013848.2 | NP_038876.2 | 11 - 12 | 1257 | 80 | |
GenBank mRNA | AF153906.1 | - | 11 - 12 | 1222 | 80 | |
AK036702.1 | - | 11 - 12 | 1218 | 80 | ||
AK158114.1 | - | 11 - 12 | 1236 | 80 | ||
BC063257.1 | - | 11 - 12 | 1191 | 6410069 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |