Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900007669.mm.1 |
NM_001136091.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007669.mm.1 |
NM_001136091.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007669.mm.1 |
NM_001167784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007669.mm.1 |
NM_001167784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007669.mm.1 |
NM_016921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007669.mm.1 |
NM_016921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007669.mm.1 |
TC1900000888.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007670.mm.1 |
NM_001136091.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007670.mm.1 |
NM_001136091.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007670.mm.1 |
NM_001167784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007670.mm.1 |
NM_001167784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007670.mm.1 |
NM_016921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007670.mm.1 |
NM_016921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007670.mm.1 |
TC1900000888.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900007671.mm.1 |
NM_001136091.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007671.mm.1 |
NM_001136091.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007671.mm.1 |
NM_001167784.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007671.mm.1 |
NM_001167784.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007671.mm.1 |
NM_016921.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007671.mm.1 |
NM_016921.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900007671.mm.1 |
TC1900000888.mm.1 |
MTA TC |